Anti-POLK Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-207EA 553 EUR
PRSI46-207
Anti-POLK Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution >1:32,000. Western Blot: Experiments gave a band at approximately 40 kDa in human liver and HepG2 extracts at 1 μg/mL. Immunohistochemistry: In paraffin embedded human small intestine shows nuclear staining of enterocytes and of most cells in the lamina propria. Recommended concentration, 5-10 μg/mL.

POLL antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

POLL antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: POLK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR