Anti-PLXND1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-202EA 420 EUR
PRSI46-202
Anti-PLXND1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 28 kDa band in human bone marrow lysates after 0.1 μg/mL antibody staining. The 28 kDa band was successfully blocked by incubation with the immunizing peptide.

Plexin D1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Plexin D1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PLXND1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR