Anti-PITX3 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-197EA 553 EUR
PRSI46-197
Anti-PITX3 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:1,000. Western Blot: Approximately 40 kDa band observed in human brain (cerebellum and cerebral cortex) lysates (calculated MW of 31.8 kDa according to NP_005020.1). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Coulon et al, J Biol Chem. 2007 Nov 9;282 (45) :33192-200.; PMID: 17848564). Recommended concentration: 0.3-1 μg/mL.

PITX3 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

PITX3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PITX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR