Anti-PIN1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-189EA 553 EUR
PRSI46-189
Anti-PIN1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:128,000. Western Blot: Approximately 17 kDa band observed in rat brain lysates (calculated MW of 18.2 kDa according to human NP_006212.1 and 18.2 kDa according to mouse NP_075860). Recommended concentration: 0.2-0.6 μg/mL.

PIN1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

PIN1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR