Anti-PDE4B Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-181EA 553 EUR
PRSI46-181
Anti-PDE4B Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:8000. Western Blot: Approximately 85 kDa band observed in mouse brain lysates (calculated MW of 83.3 kDa according to human NP_002591.2 and 82.1 kDa according to mouse NP_062814.2). In transfected HEK293 transiently expressing human PDE4B a band of approximately 85 kDa is observed. This band is not observed in the non-transfected HEK293. Recommended concentration: 0.5-1.0 μg/mL.

Phosphodiesterase 4B antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

pH ospH odiesterase 4B antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PDE4B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR