Anti-PGBD2 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-172EA 435 EUR
PRSI46-172
Anti-PGBD2 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 30 kDa band in human cerebral cortex lysates after 0.3 μg/mL antibody staining.6 kDa according to NP_001017434.1 and of 68.0 kDa according to NP_733843.1. The 30 kDa band was successfully blocked by incubation with the immunizing peptide.

PGBD2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

PGBD2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PGBD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR