Anti-PEX13 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-170EA 420 EUR
PRSI46-170
Anti-PEX13 Goat Polyclonal Antibody
Antikörper
ELISA: 1: 16,000. Western Blot: Consistent band observed in 293 at approximately 65 kDa with 1 hour incubation of primary at 1 μg/mL.

PEX13 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

PEX13 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PEX13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR