Anti-PLIN1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-168EA 553 EUR
PRSI46-168
Anti-PLIN1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 55-60 kDa band observed in human adipose lysates (calculated MW of 56.0 kDa according to NP_002657.2). Recommended concentration: 0.01-0.03 μg/mL.

Perilipin antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Perilipin antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PLIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR