Anti-PDE5A Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-153EA 553 EUR
PRSI46-153
Anti-PDE5A Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 100 kDa band observed in human lung lysates (calculated MW of 100 kDa according to isoform 1, NP_001074.2). In transfected HEK293 transiently expressing PDE5A isoform 2 a band of approximately 95 kDa is observed. This band is not observed in the non-transfected HEK293 (calculated MW of 94.8 kDa according to isoform 2, NP_236914.2). Recommended concentration: 0.3-1 μg/mL.

PDE5A antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

PDE5A antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PDE5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR