Anti-PDE4D Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-152EA 420 EUR
PRSI46-152
Anti-PDE4D Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:128,000. Western Blot: Approximately 75 kDa band observed in rat brain lysates (calculated MW of 76.5 kDa according to human aAA97892.1). Recommended concentration: 0.01-0.03 μg/mL.

PDE4D3 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

PDE4D3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PDE4D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR