Anti-PAX8 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-141EA 553 EUR
PRSI46-141
Anti-PAX8 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution >1:32,000. Western Blot: an approximately 40 kDa band observed in Human Liver lysates. Recommended concentration: 1-3 μg/mL. Immunohistochemistry: In paraffin embedded Human Thyroid Gland shows nuclear staining in epithelial cells around follicles. Recommended concentration, 1-2 μg/mL. Does not stain mouse tissues

PAX8 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

PAX8 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PAX8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR