Anti-P2RY1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-123EA 345 EUR
PRSI46-123
Anti-P2RY1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:128,000. Western Blot: Approximately 55 kDa and 38 kDa in lysates of cell lines HEK293 and A549 after 0.1 μg/mL antibody staining.1 kDa according to NP_002554.1. Both detected bands were successfully blocked by incubation with the immunizing peptide (and BLAST results with the immunizing peptide sequence did not identify any other proteins to explain the observed sizes).

P2RY1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

P2RY1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: P2RY1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR