Anti-NCAM2 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-990EA 553 EUR
PRSI45-990
Anti-NCAM2 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:4,000. Western Blot: Approximately 26 kDa band in human and mouse brain lysates after 0.1 μg/mL antibody staining.0 kDa according to NP_004531.2. The 26 kDa band was successfully blocked by incubation with the immunizing peptide.

OCAM antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

OCAM antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NCAM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR