Anti-NUP62 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-986EA 345 EUR
PRSI45-986
Anti-NUP62 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:8,000. Western Blot: Approximately 30 kDa band in human testis lysates after 0.5 μg/mL antibody staining. The 30 kDa band was successfully blocked by incubation with the immunizing peptide.

NUP62 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

NUP62 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NUP62
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR