Anti-NPY2R Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-980EA 420 EUR
PRSI45-980
Anti-NPY2R Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 40+45 kDa bands observed in mouse brain and rat brain lysates (calculated MW of 42.6 kDa according to NP032757.1). Recommended concentration: 0.1-0.3 μg/mL. Immunoprecipitation: A customer successfully used this antibody for IP of human Y2R.

Npy2r antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Npy2r antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NPY2R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR