Anti-GRIN1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-971EA 420 EUR
PRSI45-971
Anti-GRIN1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:1,000. Western Blot: Approximately 110 kDa band observed in rat brain lysates (calculated MW of 105 kDa according to NP_015566.1). Recommended concentration: 2-4 μg/mL. A minor band of unknown identity was also consistently observed at 23-24 kDa . This band was strong in mouse brain lysates and faint in human brain lysates. The band was successfully blocked by incubation with the immunizing peptide.

NMDA Receptor antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

NMDA Receptor antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GRIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR