Anti-NHEJ1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-966EA 420 EUR
PRSI45-966
Anti-NHEJ1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 38 kDa band observed in lysates of human skin, testis and thyroid gland (calculated size of 33.3 kDa according to NP_079058.1). The observed molecular weight corresponds to findings with antibodies from other sources. The 38 kDa band was successfully blocked by incubation with the immunizing peptide. Recommended concentration 0.1-0.3 μg/mL. Immunohistochemistry: In paraffin embedded human placenta shows nuclear staining in trophoblasts. Recommended concentration, 2.5-3.8 μg/mL.

NHEJ1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

NHEJ1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NHEJ1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR