Anti-NBEA Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-951EA 420 EUR
PRSI45-951
Anti-NBEA Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:8,000. Western Blot: Approximately 300 kDa band observed in lysates of transfected HEK293 and endogenously in lysates of insulinoma cell line B-TC3 (calculated MW of 327 kDa according to NP_056493.3). Data kindly provided by Professor John Creemers, K.U. Leuven, Belgium. Recommended concentration: 1-3 μg/mL.

Neurobeachin antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Neurobeachin antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NBEA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR