Anti-NENF Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-950EA 345 EUR
PRSI45-950
Anti-NENF Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 45 kDa band in human cerebellum lysates after 0.3 μg/mL antibody staining.6 kDa according to NP_037481.1 The 45 kDa band was successfully blocked by incubation with the immunizing peptide.

Neudesin antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Neudesin antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NENF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR