Anti-NCF4 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-937EA 420 EUR
PRSI45-937
Anti-NCF4 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 38 kDa band observed in lysates of Daudi lysate (calculated MW of 39.0 kDa according to NP_000622.2 and 38.8 kDa according to NP_038202.1). In transfected HEK293 transiently expressing P40PHOX a band of approximately 42 kDa is observed. This band is not observed in the non-transfected HEK293. Recommended concentration: 0.3-1 μg/mL. Immunohistochemistry: In paraffin embedded human tonsil shows staining of the cytoplasm in some cells. Recommended concentration, 2-4 μg/mL.

NCF4 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

NCF4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NCF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR