ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 85 kDa band in human muscle lysates after 0.1 μg/mL antibody staining.5 kDa according to NP_908973.1. The 85 kDa band was successfully blocked by incubation with the immunizing peptide.
MURF2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
MURF2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TRIM55
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human