Anti-MTMR9 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-912EA 553 EUR
PRSI45-912
Anti-MTMR9 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:8000. Immunohistochemistry: Immunohistochemsitry: This antibody has been successfully used on rat Hypothalamus.

MTMR9 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

MTMR9 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MTMR9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR