Anti-MLL4 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-891EA 420 EUR
PRSI45-891
Anti-MLL4 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:128000. Western Blot: Preliminary experiments gave no signal but low background in human heart lysates at up to 1 μg/mL. However this used our routine Western Blotting protocol which we would not expect to reliably detect proteins as large as the predicted size of approximately 312 kDa according to NP_055542. Immunohistochemistry: Immunohistochemistry: In paraffin embedded human testis shows cytoplasm staining of spermatids. Recommended concentration, 5-10 μg/mL.

MLL4 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

MLL4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MLL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR