Anti-SERPINI2 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-884EA 420 EUR
PRSI45-884
Anti-SERPINI2 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 40 kDa band observed in human pancreas lysates (calculated MW of 46.1 kDa according to NP_006208.1 and 47.4 kDa according to NP_001012303.1). Recommended concentration: 0.3-1 μg/mL.

MEPI antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

MEPI antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SERPINI2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR