Anti-MEPCE Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-883EA 420 EUR
PRSI45-883
Anti-MEPCE Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:2,000. Western Blot: Approximately 75 kDa band observed in rat testis lysates (calculated MW of 74.4 kDa according to human NP_062552.2 and 75.3 kDa according to rat XP_222048.4). In transfected HEK293 transiently expressing human MEPCE a band of approximately 75 kDa is observed. This band is not observed in the non-transfected HEK293. Recommended concentration: 1-3 μg/mL.

MEPCE antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

MEPCE antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MEPCE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR