Anti-MAP3K7IP3 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-863EA 345 EUR
PRSI45-863
Anti-MAP3K7IP3 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:8,000. Western Blot: Approximately 48 kDa band in human kidney lysates after 0.3 μg/mL antibody staining. The 48 kDa band was successfully blocked by incubation with the immunizing peptide.

MAP3K7IP3 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

MAP3K7IP3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MAP3K7IP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR