Anti-SCGB2A2 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-859EA 553 EUR
PRSI45-859
Anti-SCGB2A2 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Not yet tested - our routinely used Western Blotting protocol does not allow detection of proteins as small as the calculated size of 10.5 kDa according to NP_002402.1. Therefore we cannot recommend an optimal concentration and the product is investigative grade. Immunohistochemistry: In paraffin embedded human breast shows staining of cytoplasm in some acinar cells. Recommended concentration, 4-6 μg/mL.

Mammaglobin A antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Mammaglobin A antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SCGB2A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR