Anti-MXD4 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-855EA 420 EUR
PRSI45-855
Anti-MXD4 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 22 kDa band observed in human kidney lysates (predicted MW of 24 kDa according to NP_006445). Recommended for use at 0.5-2 μg/mL. Please note the appearance of this band was blocked by incubation with the immunizing peptide.

MAD4 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

MAD4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MXD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR