Anti-LRRK2 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-848EA 553 EUR
PRSI45-848
Anti-LRRK2 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64,000. Antibody has been successfully used in both Western Blot and immunohistochemistry on human brain: Alegre-Abarrategui J et al., Neuropathol Appl Neurobiol. 2007 Oct 26; PMID: 17971075. Recommended concentration: 0.3-1 μg/mL. Immunohistochemistry: In paraffin embedded human hippocampus shows staining of neuronal cytoplasm. Recommended concentration, 1-2 μg/mL.

LRRK2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

LRRK2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: LRRK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR