Anti-CDKN1B Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-800EA 420 EUR
PRSI45-800
Anti-CDKN1B Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 26 kDa band observed in lysates of human prostate and of hepatoblastoma cell line HepG2 (calculated MW of 22.1 kDa according to NP_004055.1). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Xia et al, Proc Natl Acad Sci U S A. 2005 Sep 27;102 (39) :14028-33; PMID: 16169901). Recommended concentration: 0.1-0.3 μg/mL.

KIP1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

KIP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CDKN1B
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR