Anti-KALRN Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-789EA 553 EUR
PRSI45-789
Anti-KALRN Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:4,000. Western Blot: Approximately 200 kDa band observed in mouse brain lysates (calculated MW of 192 kDa according to XP_001001845). Recommended concentration: 1-3 μg/mL.

Kalirin antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Kalirin antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: KALRN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR