Anti-MAPK8IP3 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-788EA 420 EUR
PRSI45-788
Anti-MAPK8IP3 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:4,000. Immunohistochemistry: In paraffin embedded mouse brain shows axonal staining in various regions. Recommended concentration, 4-6 μg/mL.

JIP3 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

JIP3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MAPK8IP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR