Anti-ITGAM Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-783EA 420 EUR
PRSI45-783
Anti-ITGAM Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Approximately 170 kDa band observed in human cerebellum lysates (calculated MW of 127 kDa according to NP_000623.2). The observed molecular weight corresponds to earlier findings with different antibodies from other sources. Recommended concentration: 1-3 μg/mL.

ITGAM antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ITGAM antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ITGAM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR