Anti-ITGB5 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-777EA 553 EUR
PRSI45-777
Anti-ITGB5 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 110 kDa band observed in human ovary and mouse eye lysates (calculated MW of 88.1 kDa according to NP_002204.2). Recommended concentration: 0.1-0.3 μg/mL. Customer feedback: observed 110 kDa in mouse eye was confirmed and not present in knock-out mouse.

Integrin beta 5 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Integrin beta 5 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ITGB5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR