Anti-l12b Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-766EA 420 EUR
PRSI45-766
Anti-l12b Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:4,000. Western Blot: Approximately 37 kDa band observed in mouse spleen and rat liver lysates (calculated MW of 38.2 kDa according to mouse NP_032378.1 and 38.4 kDa according to rat NP_072133.1). Recommended concentration: 1-3 μg/mL.

Il12 beta antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Il12 beta antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: l12b
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR