Anti-IER5 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-761EA 420 EUR
PRSI45-761
Anti-IER5 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:2000. Western Blot: Customer's experiments gave a band at approximately 45 kDa in lysates of transfected H1299 overexpressing IER5 but not when transfected with empty vector (calculated MW of 33.7 kDa according to NP_057629.1). Recommended concentration: 1-3 μg/mL.

IER5 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

IER5 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: IER5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR