Anti-HTATSF1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-752EA 420 EUR
PRSI45-752
Anti-HTATSF1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 140 kDa band observed in lysates of Daudi lysate (calculated MW of 85.9 kDa according to NP_055315.2). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Zhou and Sharp, Science. 1996 Oct 25;274 (5287) :605-10.; PMID: 8849451). Recommended concentration: 1-3 μg/mL. an additional band of unknown identity was also consistently observed at 20 kDa . This band was successfully blocked by incubation with the immunizing peptide.

HTATSF1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

HTATSF1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: HTATSF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR