Anti-HRNR Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-743EA 345 EUR
PRSI45-743
Anti-HRNR Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 50 kDa band in human skin lysates after 0.3 μg/mL antibody staining.1. The 50 kDa band was successfully blocked by incubation with the immunizing peptide.

Hornerin antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Hornerin antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: HRNR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR