Anti-HRH1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-731EA 553 EUR
PRSI45-731
Anti-HRH1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 55 kDa and 37 kDa in human duodenum lysate after 0.3 μg/mL antibody staining.8 kDa according to NP_000852.1. Both detected bands were successfully blocked by incubation with the immunizing peptide (and BLAST results with the immunizing peptide sequence did not identify any other proteins to explain the additional band). Immunohistochemistry: In paraffin embedded human colon shows membranous staining. Recommended concentration, 4-6 μg/mL.

Histamine Receptor H1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Histamine Receptor H1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: HRH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR