Anti-DBNL Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-730EA 420 EUR
PRSI45-730
Anti-DBNL Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1/32,000. Western Blot: Approximately 55 kDa band observed in Jurkat and 3T3 lysates (predicted MW of 49 kDa according to NP_054782). Recommended for use at 1-3 μg/mL.

HIP-55 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

HIP-55 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: DBNL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR