Anti-Hap1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-722EA 553 EUR
PRSI45-722
Anti-Hap1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 110 kDa band in mouse and rat brain lysates after 0.5 μg/mL antibody staining.8 kDa according to NP_034534.1. The 110 kDa band was successfully blocked by incubation with the immunizing peptide.

HAP1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

HAP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: Hap1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR