Anti-ARHGAP26 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-703EA 553 EUR
PRSI45-703
Anti-ARHGAP26 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 90 kDa band observed in human heart lysates (calculated MW of 92.2 kDa according to NP_055886.1). Recommended concentration: 0.3-1 μg/mL. an additional strong band of unknown identity was also consistently observed at 16 kDa . This band was successfully blocked by incubation with the immunizing peptide.

GRAF antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

GRAF antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ARHGAP26
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR