Anti-GPX3 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-699EA 553 EUR
PRSI45-699
Anti-GPX3 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:1,000. Western Blot: Approximately 26 kDa band observed in mouse kidney and liver lysates (calculated MW of 27.7 kDa according to mouse NP_001077398.1 [isoform1], 25.4 kDa according to mouse NP_032187.2 [isoform 2] and 25.5 kDa according to human NP_002075.2). Recommended concentration: 1-3 μg/mL.

GPX3 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

GPX3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GPX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR