Anti-GPSM2 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-698EA 553 EUR
PRSI45-698
Anti-GPSM2 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 70 kDa band observed in human cerebellum and human liver lysates (calculated MW of 75.8 kDa according to NP_037428.2). Recommended concentration: 0.01-0.03 μg/mL. an additional band of unknown identity was also consistently observed at 26-28 kDa . This band was successfully blocked by incubation with the immunizing peptide.

GPSM2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

GPSM2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GPSM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR