Anti-GOT1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-690EA 420 EUR
PRSI45-690
Anti-GOT1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:4000. Western Blot: Approximately 38-40 kDa band observed in human cerebellum, liver and skeletal muscle lysates (calculated MW of 46.2 kDa according to NP_002070.1). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Berk et al, Proc Natl Acad Sci U S A. 1990 May;87 (9) :3484-8; PMID: 2185471). Recommended concentration: 0.01-1 μg/mL. Immunohistochemistry: In paraffin embedded human heart shows strong staining of the capillary network between the myocardial fibres in transverse secion. Recommended concentration, 2-3 μg/mL.

GOT1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

GOT1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GOT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR