Anti-GOLGA6B Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-687EA 420 EUR
PRSI45-687
Anti-GOLGA6B Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:1,000. Western Blot: Approximately 45 kDa band in human liver lysates after 1 μg/mL antibody staining. The 45 kDa band was successfully blocked by incubation with the immunizing peptide.

GOLGA6 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

GOLGA6 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GOLGA6B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR