Anti-GOLGA3 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-686EA 420 EUR
PRSI45-686
Anti-GOLGA3 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 170 kDa band observed in human HeLa lysates (calculated MW of 167 kDa according to NP_005886.2). Recommended concentration: 0.1-0.3 μg/mL. an additional band of unknown identity was also consistently observed at appox 25 kDa . This band was successfully blocked by incubation with the immunizing peptide.

GOLGA3 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

GOLGA3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GOLGA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR