Anti-SLC2A8 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-680EA 345 EUR
PRSI45-680
Anti-SLC2A8 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 80 kDa and 35 kDa in human peripheral blood mononucleocyte lysates after 0.5 μg/mL antibody staining.8 kDa according to NP_055395.2. Both detected bands were successfully blocked by incubation with the immunizing peptide (and BLAST results with the immunizing peptide sequence did not identify any other proteins to explain the additional bands). Added note: The observed 35 kDa band corresponds to the calculated size of 34.4 kDa of EAW87663.1 (314 aa long).

GLUT8 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

GLUT8 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SLC2A8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR