Anti-GLTSCR2 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-677EA 345 EUR
PRSI45-677
Anti-GLTSCR2 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 16-18 kDa band in human heart, liver and breast tumor lysates after 0.2 μg/mL antibody staining.4 kDa according to NP_056525.1. The 16-18 kDa band was successfully blocked by incubation with the immunizing peptide.

GLTSCR2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

GLTSCR2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GLTSCR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR