Anti-GLI2 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-674EA 420 EUR
PRSI45-674
Anti-GLI2 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:2,000. Western Blot: Approximately 55 kDa band in human bone marrow and ovary lysates after 1 μg/mL antibody staining.2. The 55 kDa band was successfully blocked by incubation with the immunizing peptide.

GLI2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

GLI2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GLI2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR